1-Piperidinecarbonyl chloride [carbonyl-14C] ≥98% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC2306-50UCI
101971-380EA 5191.54 USD
101971-380
1-Piperidinecarbonyl chloride [carbonyl-14C] ≥98% (by HPLC), MORpure™
1-Piperidinecarbonyl chloride [carbonyl-14C]
CAS Number: 188045-82-1
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR