Fluroxypyr[methylene-14C]-1-methylheptyl ester ≥98% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC2291-50UCI
101971-346EA 5191.54 USD
101971-346
Fluroxypyr[methylene-14C]-1-methylheptyl ester ≥98% (by HPLC), MORpure™
Fluroxypyr[methylene-14C]-1-methylheptyl ester
CAS Number: 2088617-20-1
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR