Thymine, [2,6-14C] ≥98% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC2237-10UCI
101971-244EA 1621.54 USD
101971-244
Thymine, [2,6-14C] ≥98% (by HPLC), MORpure™
Thymine, [2,6-14C]
CAS Number: 90980-11-3
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR