9-(2-Phosphonylmethoxyethyl)guanine, [guanine-8-14C] ≥98% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC2231-10UCI
101971-228EA 1621.54 USD
101971-228
9-(2-Phosphonylmethoxyethyl)guanine, [guanine-8-14C] ≥98% (by HPLC), MORpure™
9-(2-Phosphonylmethoxyethyl)guanine, [guanine-8-14C]
CAS Number: 151842-62-5
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR