3-Fluoropyridine, [2-14C] ≥98% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC2218-50UCI
101971-206EA 5191.54 USD
101971-206
3-Fluoropyridine, [2-14C] ≥98% (by HPLC), MORpure™
3-Fluoropyridine, [2-14C]
CAS Number: 2088577-49-3
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR