1-Nitronaphthalene [14C] ≥98% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC2195-50UCI
101971-138EA 5191.54 USD
101971-138
1-Nitronaphthalene [14C] ≥98% (by HPLC), MORpure™
1-Nitronaphthalene [14C]
CAS Number: 2088206-35-1
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR