Benadryl, [phenyl-14C] ≥98% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC2158-50UCI
101971-096EA 5191.54 USD
101971-096
Benadryl, [phenyl-14C] ≥98% (by HPLC), MORpure™
Benadryl, [phenyl-14C]
CAS Number: 2088206-03-3
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR