L-Thymidine, [2-14C] ≥98% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC2096-50UCI
101971-024EA 5191.54 USD
101971-024
L-Thymidine, [2-14C] ≥98% (by HPLC), MORpure™
L-Thymidine, [2-14C]
CAS Number: 2089095-31-6
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR