2-Methoxy-4-amino-5-chlorobenzoic acid, [carboxy-14C] ≥98% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC2078-50UCI
101971-008EA 5191.54 USD
101971-008
2-Methoxy-4-amino-5-chlorobenzoic acid, [carboxy-14C] ≥98% (by HPLC), MORpure™
2-Methoxy-4-amino-5-chlorobenzoic acid, [carboxy-14C]
CAS Number: 116277-20-4
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR