(-)-β-2',3'-Dideoxy-3'-thia-5-fluorocytidine, [2-14C] ≥98% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC2031-10UCI MC2031-50UCI
101970-918EA 1621.54 USD
101970-918 101970-920
(-)-β-2',3'-Dideoxy-3'-thia-5-fluorocytidine, [2-14C] ≥98% (by HPLC), MORpure™
(-)-β-2',3'-Dideoxy-3'-thia-5-fluorocytidine, [2-14C]
CAS Number: 2088208-87-9
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR