D-Mannitol, [1-14C] ≥98% (by HPLC), MORpure™

MC203-50UCI MC203-250UCI
101970-916EA 1598.05 USD
101970-916 101970-914
D-Mannitol, [1-14C] ≥98% (by HPLC), MORpure™
D-Mannitol, [1-14C]
CAS Number: 88404-24-4
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR