Guanosine-5'-triphosphate, tetraammonium salt, [8-14C] ≥97% (by HPLC)

MC194-50UCI
101970-840EA 4375.34 USD
101970-840
Guanosine-5'-triphosphate, tetraammonium salt, [8-14C] ≥97% (by HPLC)
Guanosine-5'-triphosphate, tetraammonium salt, [8-14C]
CAS Number: 335021-94-8
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR