Hypoxanthine, [14C(U)] ≥98% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC167-10UCI
101970-746EA 1621.54 USD
101970-746
Hypoxanthine, [14C(U)] ≥98% (by HPLC), MORpure™
Hypoxanthine, [14C(U)]
CAS Number: 2088577-51-7
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR