Guanine hydrochloride, [8-14C] ≥98% (by HPLC), MORpure™

MC162-50UCI MC162-250UCI
101970-738EA 1945.21 USD
101970-738 101970-734
Guanine hydrochloride, [8-14C] ≥98% (by HPLC), MORpure™
Guanine hydrochloride, [8-14C]
CAS Number: 98853-66-8
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR