Cyanoguanidine, [14C(U)] ≥98% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC1484-1MCI
101970-672EA 5283.08 USD
101970-672
Cyanoguanidine, [14C(U)] ≥98% (by HPLC), MORpure™
Cyanoguanidine, [14C(U)]
CAS Number: 3403-66-5
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR