Phenylacetic acid, [acetate-1,2-14C] ≥98% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC1449-250UCI
101970-594EA 1621.54 USD
101970-594
Phenylacetic acid, [acetate-1,2-14C] ≥98% (by HPLC), MORpure™
Phenylacetic acid, [acetate-1,2-14C]
CAS Number: 289906-86-1
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR