2,4,6-Trinitrotoluene, [methyl-14C] ≥97% (by HPLC)

Supplier: MORAVEK BIOCHEMICALS MS

MC1441-50UCI
101970-574EA 5191.54 USD
101970-574
2,4,6-Trinitrotoluene, [methyl-14C] ≥97% (by HPLC)
2,4,6-Trinitrotoluene, [methyl-14C]
CAS Number: 91746-63-3
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR