D-Glucose, [14C(U)] ≥98% (by HPLC), MORpure™

MC144-1MCI MC144-50UCI MC144-250UCI
101970-570EA 1449.26 USD
101970-570 101970-572 101970-568
D-Glucose, [14C(U)] ≥98% (by HPLC), MORpure™
D-Glucose, [14C(U)]
CAS Number: 2088920-79-8
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR