Thiourea, [14C] ≥98% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC140-5MCI
101970-484EA 5368.08 USD
101970-484
Thiourea, [14C] ≥98% (by HPLC), MORpure™
Thiourea, [14C]
CAS Number: 5022-68-4
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR