p-Fluorobenzoic acid, [7-14C] ≥98% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC1398-50UCI
101970-472EA 5191.54 USD
101970-472
p-Fluorobenzoic acid, [7-14C] ≥98% (by HPLC), MORpure™
p-Fluorobenzoic acid, [7-14C]
CAS Number: 1523-98-4
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR