Cytidine-5'-monophosphate, diammonium salt [2-14C] ≥97% (by HPLC)

Supplier: MORAVEK BIOCHEMICALS MS

MC1379-10UCI
101970-404EA 1771.92 USD
101970-404
Cytidine-5'-monophosphate, diammonium salt [2-14C] ≥97% (by HPLC)
Cytidine-5'-monophosphate, diammonium salt [2-14C]
CAS Number: 130150-79-7
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR