Formaldehyde, [14C] 1 - 3% in aqueous solution ≥ 20 mCi (740 MBq)/mmol
|
|
MC1307H-1MCI
MC1307H-250UCI
MC1307H-50UCI
101970-320EA
451.86
USD
101970-320
101970-322
101970-324
Formaldehyde, [14C] 1 - 3% in aqueous solution ≥ 20 mCi (740 MBq)/mmol
Formaldehyde, [14C]
Storage Temperature:
Refrigerator |
CAS Number:
3046-49-9 UN: 2910 ADR: , |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...