Formaldehyde, [14C] 1% in aqueous solution, 2-10 mCi 74-370 MBq/mmol

MC1307-1MCI
101970-318EA 2237.26 USD
101970-318
Formaldehyde, [14C] 1% in aqueous solution, 2-10 mCi 74-370 MBq/mmol
Formaldehyde, [14C]
Storage Temperature: Refrigerator
CAS Number: 3046-49-9
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR