Chloroacetic acid, [2-14C] ≥97% (by HPLC)

MC1264-250UCI
101970-290EA 1961.74 USD
101970-290
Chloroacetic acid, [2-14C] ≥97% (by HPLC)
Chloroacetic acid, [2-14C]
CAS Number: 2946-86-3
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR