Acetone-[2-14C]

MC1231-250UCI MC1231-1MCI
101970-232EA 1901.12 USD
101970-232 101970-234
Acetone-[2-14C]
Acetone-[2-14C]
Formula: C₃H₆O
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR