Acetaminophen, [ring-14C(U)] ≥98% (by HPLC), MORpure™

MC1227-100UCI MC1227-250UCI
101970-218EA 42137.31 USD
101970-218 101970-216
Acetaminophen, [ring-14C(U)] ≥98% (by HPLC), MORpure™
Acetaminophen, [ring-14C(U)]
CAS Number: 1313828-45-3
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR