Phenanthrene, [9-14C] ≥98% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC1178-100UCI MC1178-250UCI
101970-178EA 2981.54 USD
101970-178 101970-180
Phenanthrene, [9-14C] ≥98% (by HPLC), MORpure™
Phenanthrene, [9-14C]
CAS Number: 97193-05-0
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR