L-Carnitine hydrochloride, [(N)methyl-14C] ≥97% (by HPLC)

Supplier: MORAVEK BIOCHEMICALS MS

MC1147-50UCI MC1147-10UCI
101970-144EA 1621.54 USD
101970-144 101970-146
L-Carnitine hydrochloride, [(N)methyl-14C] ≥97% (by HPLC)
L-Carnitine hydrochloride, [(N)methyl-14C]
CAS Number: 2088236-24-0
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR