L-Glutamine, [14C(U)] ≥97% (by HPLC)

Supplier: MORAVEK BIOCHEMICALS MS

MC1124-50UCI MC1124-250UCI
101970-124EA 5191.54 USD
101970-124 101970-126
L-Glutamine, [14C(U)] ≥97% (by HPLC)
L-Glutamine, [14C(U)]
CAS Number: 152154-51-3
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR