Glycine, [1-14C] ≥98% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC1109-250UCI MC1109-50UCI MC1109-1MCI
101970-110EA 6885 USD
101970-110 101970-108 101970-106
Glycine, [1-14C] ≥98% (by HPLC), MORpure™
Glycine, [1-14C]
CAS Number: 56-39-3
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR