Hydroquinone, [ring-14C] ≥98% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC1101-250UCI
101970-088EA 6120 USD
101970-088
Hydroquinone, [ring-14C] ≥98% (by HPLC), MORpure™
Hydroquinone, [ring-14C]
CAS Number: 106817-54-3
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR