β-L-Adenosine ≥98% (by HPLC) MORpure™
Supplier: MORAVEK BIOCHEMICALS MS
Synonyms:
(2S,3S,4R,5S)-2-(6-Amino-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol
M530-1MG
M530-5MG
101969-942EA
738.85
USD
101969-942
101969-940
β-L-Adenosine ≥98% (by HPLC) MORpure™
β-L-Adenosine
Formula:
C₁₀H₁₃N₅O₄ MW: 267.2413 g/mol |
MDL Number:
MFCD01075784 CAS Number: 3080-29-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...