β-L-Uridine ≥98% (by HPLC) MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

Synonyms: 1-((2S,3S,4R,5S)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione

M321-5MG M321-1MG
101969-892EA 281.15 USD
101969-892 101969-894
β-L-Uridine ≥98% (by HPLC) MORpure™
β-L-Uridine
Formula: C₉H₁₂N₂O₆
MW: 244.2 g/mol
MDL Number: MFCD02683606
CAS Number: 26287-69-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR