L(+)-Valine ≥95%

Supplier: MORAVEK BIOCHEMICALS MS

Synonyms: (S)-(+)-ValineValineH-Val-OH

M1918-1MG M1918-5MG
101969-868EA 738.85 USD
101969-868 101969-866
L(+)-Valine ≥95%
L(+)-Valine
Formula: C₅H₁₁NO₂
MW: 117.15 g/mol
MDL Number: MFCD00064220
CAS Number: 72-18-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR