Etidronate disodium ≥98% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

Synonyms: Dihydrogen (1-hydroxyethylidene)bisphosphonate disodiumDisodium hydrogen (1-hydrogen phosphonato-1-hydroxyethyl)phosphonate

M1831-5MG M1831-1MG
101969-746EA 738.85 USD
101969-746 101969-744
Etidronate disodium ≥98% (by HPLC), MORpure™
Etidronate disodium
Formula: C₂H₆Na₂O₇P₂
MW: 249.99 g/mol
MDL Number: MFCD00152567
CAS Number: 1159812-97-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR