2'-O-Methylcytidine
Supplier: MORAVEK BIOCHEMICALS MS
M1813-1MG
M1813-5MG
101969-690EA
738.85
USD
101969-690
101969-688
2'-O-Methylcytidine
2'-O-Methylcytidine
Formula:
C₁₀H₁₅N₃O₅ MW: 257.25 g/mol |
MDL Number:
MFCD00056067 CAS Number: 2140-72-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...