3'-(O-Methyl)-guanosine ≥98% (by HPLC) MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

Synonyms: 2-Amino-9-((2R,3R,4S,5R)-3-hydroxy-5-(hydroxymethyl)-4-methoxytetrahydrofuran-2-yl)-1,9-dihydro-6H-purin-6-one

M1743-1MG
101969-432EA 281.15 USD
101969-432
3'-(O-Methyl)-guanosine ≥98% (by HPLC) MORpure™
3'-(O-Methyl)guanosine
Formula: C₁₁H₁₅N₅O₅
MW: 297.2673 g/mol
MDL Number: MFCD00057057
CAS Number: 10300-27-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR