Amprenavir ≥98% (by HPLC) MORpure™
Supplier: MORAVEK BIOCHEMICALS MS
Synonyms:
(S)-Tetrahydrofuran-3-yl ((2S,3R)-4-((4-amino-N-isobutylphenyl)sulphonamido)-3-hydroxy-1-phenylbutan-2-yl)carbamate
, (S)-Tetrahydrofuran-3-yl ((2S,3R)-4-((4-amino-N-isobutylphenyl)sulfonamido)-3-hydroxy-1-phenylbutan-2-yl)carbamate
M1613-1MG
M1613-5MG
M1613-10MG
101969-326EA
980.77
USD
101969-326
101969-324
101969-322
Amprenavir ≥98% (by HPLC) MORpure™
Amprenavir
Formula:
C₂₅H₃₅N₃O₆S MW: 505.6269 g/mol |
MDL Number:
MFCD00934214 CAS Number: 161814-49-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...