Emtricitabine triphosphate, tetraammonium salt ≥93% (by HPLC)

M1596-2UMOL
101969-296EA 4171.45 USD
101969-296
Emtricitabine triphosphate, tetraammonium salt ≥93% (by HPLC)
Emtricitabine triphosphate, tetraammonium salt
Storage Temperature: Freezer
CAS Number: 305813-63-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR