SP Wilmad-LabGlass Ground Flat Flange Joints, SP Industries
Supplier: SP Industries
LG-1023-104 LG-1023-108 LG-1023-106 LG-1023-102 LG-1023-100
14212-798EA 105.25 USD
14212-798 14205-698 14202-938 14203-396 14208-368
SP Wilmad-LabGlass Ground Flat Flange Joints, SP Industries
The mating end of these borosilicate glass joints have a flat flange with a finely ground face for a liquid light seal.
Overall length is approximately 20.3cm (8"). For use with horseshoe shaped clamp (LG-7316), sold separately.