Would you like to log in again?
VWR® Flammable Storage Cabinets
Supplier: VWR International
VWR®
G65M4418430009 G65M441843009V G65M651843009V
Storage cabinets feature 18 gauge double wall construction with 11/2" air space
  • Two grounding attachment points in rear
  • Electrostatically applied powder coat finish
Upper and Lower diametrically opposed side vents.
Certifications: FM Approved.
ORDER
Expand All / Collapse All
Description Capacity Depth Height Width Color Door Type Label Type No. of Shelves VWR Catalog Number Unit Price Quantity
Flammable Storage Cabinet 30 Gallon 18" 44" 43" Yellow Self Close Flammable 1 97044-206 Each Retrieving
Product available on GSA Advantage®
Flammable Storage Cabinet 30 Gallon 18" 44" 43" Yellow Manual Close Flammable 1 97044-198 Each Retrieving
Product available on GSA Advantage®
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty