Anti-Siglec-H Rat Monoclonal Antibody [clone: 440c]

Supplier: BioXCell
BE0202-25MG BE0202-100MG BE0202-50MG BE0202-50MG BE0202-5MG BE0202-25MG BE0202-1MG BE0202-100MG BE0202-1MG BE0202-5MG
103253-914EA 550 USD
103253-914 76555-534 103253-912 103253-918 76554-680 103253-916 76557-068 76556-964 103572-910 76557-192
Anti-Siglec-H Rat Monoclonal Antibody [clone: 440c]
Antibodies
InVivoMab anti-mouse Siglec-H, Clone: 440c, Isotype: rat IgG2b, Applications: in vivo administration, Flow cytometry

Type: Primary
Antigen: Siglec-H
Clonality: Monoclonal
Clone: 440c
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR