Anti-NKG2D Armenian Hamster Monoclonal Antibody [clone: HMG2D]

Supplier: BioXCell
BE0111-1MG BE0111-50MG BE0111-5MG BE0111-100MG BE0111-100MG BE0111-50MG BE0111-25MG BE0111-5MG BE0111-25MG BE0111-1MG
103572-528EA 150 USD
103572-528 103253-714 76556-610 103253-708 76556-572 76555-608 76554-464 76555-070 103253-712 103253-710
Anti-NKG2D Armenian Hamster Monoclonal Antibody [clone: HMG2D]
Antibodies
InVivoMab anti-mouse NKG2D, Clone: HMG2D, Isotype: Armenian hamster IgG, Applications: in vivo NKG2D blockade

Type: Primary
Antigen: NKG2D
Clonality: Monoclonal
Clone: HMG2D
Conjugation:
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR