Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TCR ?/? Armenian Hamster Monoclonal Antibody [clone: UC7-13D5]

Supplier: Bio X Cell
BE0070-R001MG BE0070-R050MG BE0070-R100MG BE0070-R025MG BE0070-R005MG
103572-760 103253-644 103253-638 103253-642 103253-640
Anti-TCR ?/? Armenian Hamster Monoclonal Antibody [clone: UC7-13D5]
InVivoMab anti-mouse TCR γ/δ, Clone: UC7-13D5, Isotype: Armenian hamster IgG, κ, Applications: in vivo TCR γ/δ neutralization, in vitro γ/δ T cell stimulation, in vitro γ/δ T cell purification, Functional assays, Immunoprecipitation, Flow cytometry

Type: Primary
Antigen: TCR γ/δ
Clonality: Monoclonal
Clone: UC7-13D5
Conjugation:
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Mouse

Order Now


 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty