Anti-CD8 alpha Rat Monoclonal Antibody [clone: 2.43]

Supplier: BioXCell
BE0061-50MG BE0061-1MG BE0061-100MG BE0061-5MG BE0061-5MG BE0061-1MG BE0061-100MG BE0061-50MG BE0061-25MG BE0061-25MG
76557-012EA 3126.56 USD
76557-012 103253-580 76557-178 103253-576 76557-238 103572-496 103253-574 76557-302 103253-578 76557-208
Anti-CD8 alpha Rat Monoclonal Antibody [clone: 2.43]
Antibodies
InVivoMab anti-mouse CD8α, Clone: 2.43, Isotype: rat IgG2b, κ, Applications: in vivo CD8+ T cell depletion

Type: Primary
Antigen: CD8alpha
Clonality: Monoclonal
Clone: 2.43
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR