Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-VEGFR-2 Rat Monoclonal Antibody [clone: DC101]

Supplier: Bio X Cell
BE0060-R050MG BE0060-R005MG BE0060-R025MG BE0060-R001MG BE0060-R100MG
103253-572 103253-568 103253-570 103572-494 103253-566
Anti-VEGFR-2 Rat Monoclonal Antibody [clone: DC101]
InVivoMab anti-mouse VEGFR-2, Clone: DC101, Isotype: rat IgG1, κ, Applications: in vivo blocking of VEGF/VEGFR-2 signaling, in vitro blocking of VEGFR signaling

Type: Primary
Antigen: VEGFR-2
Clonality: Monoclonal
Clone: DC101
Conjugation:
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse

Order Now


 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty