Anti-TNFalpha Rat Monoclonal Antibody [clone: XT3.11]

Supplier: BioXCell
BE0058-100MG BE0058-1MG BE0058-50MG BE0058-5MG BE0058-1MG BE0058-100MG BE0058-25MG BE0058-25MG BE0058-50MG BE0058-5MG
103572-746EA 150 USD
103572-746 76556-654 103253-550 76555-250 103253-554 76556-094 76557-214 103253-552 76557-138 103253-556
Anti-TNFalpha Rat Monoclonal Antibody [clone: XT3.11]
Antibodies
InVivoMab anti-mouse TNFα, Clone: XT3.11, Isotype: rat IgG1, Applications: in vivo TNFα neutralization, in vitro TNFα neutralization

Type: Primary
Antigen: TNFalpha
Clonality: Monoclonal
Clone: XT3.11
Conjugation:
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR