Anti-IL-12 Rat Monoclonal Antibody [clone: R1-5D9]

Supplier: BioXCell
BE0052-100MG BE0052-5MG BE0052-25MG BE0052-100MG BE0052-1MG BE0052-25MG BE0052-5MG BE0052-50MG BE0052-50MG BE0052-1MG
103253-518EA 3920 USD
103253-518 76555-038 103572-738 76556-104 76556-592 103253-520 76554-664 103253-524 76557-070 103253-522
Anti-IL-12 Rat Monoclonal Antibody [clone: R1-5D9]
Antibodies
InVivoMab anti-mouse IL-12, Clone: R1-5D9, Isotype: rat IgG2a, Applications: in vitro IL-12 neutralization

Type: Primary
Antigen: IL-12
Clonality: Monoclonal
Clone: R1-5D9
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR