Anti-c-myc Mouse Monoclonal Antibody [clone: 9E10]

Supplier: BioXCell
RT0263-1MG RT0263-25MG RT0263-1MG RT0263-R010MG RT0263-R005MG RT0263-25MG
103253-340EA 1650 USD
103253-340 76556-782 76555-182 103253-342 103253-338 103253-336
Anti-c-myc Mouse Monoclonal Antibody [clone: 9E10]
Antibodies
ReadyTag anti-c-myc, Clone: 9E10, Isotype: mouse IgG1, Applications: Western blot, ELISA, Flow cytometry

Type: Primary
Antigen: c-myc
Clonality: Monoclonal
Clone: 9E10
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR