Anti-CD80 Mouse Monoclonal Antibody [clone: 3H5]

Supplier: BioXCell
BE0187-25MG BE0187-5MG BE0187-25MG BE0187-100MG BE0187-50MG BE0187-1MG BE0187-5MG BE0187-1MG BE0187-50MG BE0187-100MG
76555-542EA 2077.41 USD
76555-542 76555-078 103253-000 103253-004 103572-884 103253-002 76556-316 103253-006 76555-634 76556-070
Anti-CD80 Mouse Monoclonal Antibody [clone: 3H5]
Antibodies
InVivoMab anti-rat CD80, Clone: 3H5, Isotype: mouse IgG1, κ, Applications: Flow cytometry

Type: Primary
Antigen: CD80
Clonality: Monoclonal
Clone: 3H5
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR